Imprimer

Anti-HIST3H3 Mouse Monoclonal Antibody [clone: 11D8]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA328045EA 815 EUR
ORIGTA328045
Anti-HIST3H3 Mouse Monoclonal Antibody [clone: 11D8]
Anticorps
Type: Primary
Antigen: HIST3H3
Clonality: Monoclonal
Clone: 11D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR