Imprimer

Anti-SPATA1 Rabbit Polyclonal Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326615EA 0 EUR
ORIGTA326615
Anti-SPATA1 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunochemistry: 2.5 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: SPATA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR