Imprimer

Anti-VNN3 Rabbit Polyclonal Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326633EA 576 EUR
ORIGTA326633
Anti-VNN3 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: VNN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR