Imprimer

Anti-ULK4 Rabbit Polyclonal Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326686EA 618 EUR
ORIGTA326686
Anti-ULK4 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: ULK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR