Imprimer

Anti-NOS2 Rabbit Polyclonal Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA325739EA 525 EUR
ORIGTA325739
Anti-NOS2 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunochemistry: 1:50-1:200

Type: Primary
Antigen: NOS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR