Imprimer

Anti-KLF4 Mouse Monoclonal Antibody [clone: 4E5C3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326715EA 576 EUR
ORIGTA326715
Anti-KLF4 Mouse Monoclonal Antibody [clone: 4E5C3]
Anticorps
Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: KLF4
Clonality: Monoclonal
Clone: 4E5C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR