Imprimer

Anti-CR2 Mouse Monoclonal Antibody [clone: 2G9]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA327627EA 913 EUR
ORIGTA327627
Anti-CR2 Mouse Monoclonal Antibody [clone: 2G9]
Anticorps
Recommended Dilutions: Immunochemistry: 1:25-1:100

Type: Primary
Antigen: CR2
Clonality: Monoclonal
Clone: 2G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR