Imprimer

Anti-PECAM1 Mouse Monoclonal Antibody [clone: JC70]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA327635EA 761 EUR
ORIGTA327635
Anti-PECAM1 Mouse Monoclonal Antibody [clone: JC70]
Anticorps
Recommended Dilutions: Immunochemistry: 1:25-1:100

Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone: JC70
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR