Imprimer

Anti-Cd34 Mouse Monoclonal Antibody [clone: QBEnd/10]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA327637EA 1250 EUR
ORIGTA327637
Anti-Cd34 Mouse Monoclonal Antibody [clone: QBEnd/10]
Anticorps
Recommended Dilutions: Immunochemistry: 1:50-1:200

Type: Primary
Antigen: Cd34
Clonality: Monoclonal
Clone: QBEnd/10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR