Imprimer
Anti-CD44 Mouse Monoclonal Antibody [clone: MRQ-13]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA327640EA 814 EUR
ORIGTA327640
Anti-CD44 Mouse Monoclonal Antibody [clone: MRQ-13]
Anticorps
Recommended Dilutions: Immunochemistry: 1:100-1:500
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: MRQ-13
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human