Imprimer

Anti-PTPRC Mouse Monoclonal Antibody [clone: UCHL-1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA327643EA 580 EUR
ORIGTA327643
Anti-PTPRC Mouse Monoclonal Antibody [clone: UCHL-1]
Anticorps
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: UCHL-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR