Imprimer

Anti-PAK1 Rabbit Polyclonal Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326169EA 506 EUR
ORIGTA326169
Anti-PAK1 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunochemistry: 1:50-1:100 ELISA: 1:5000

Type: Primary
Antigen: PAK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR