Imprimer

Anti-SOX9 Rabbit Polyclonal Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA325869EA 564 EUR
ORIGTA325869
Anti-SOX9 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200; Immunofluorescence: 1:100-1:500

Type: Primary
Antigen: SOX9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR