Imprimer

Anti-ITGB3 Mouse Monoclonal Antibody [clone: 2f2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA327648EA 979 EUR
ORIGTA327648
Anti-ITGB3 Mouse Monoclonal Antibody [clone: 2f2]
Anticorps
Recommended Dilutions: Immunochemistry: 1:25-1:100

Type: Primary
Antigen: ITGB3
Clonality: Monoclonal
Clone: 2f2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR