Imprimer

Anti-CD74 Mouse Monoclonal Antibody [clone: LN2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA327653EA 760 EUR
ORIGTA327653
Anti-CD74 Mouse Monoclonal Antibody [clone: LN2]
Anticorps
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: CD74
Clonality: Monoclonal
Clone: LN2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR