Imprimer

Anti-CD79A Mouse Monoclonal Antibody [clone: JCB117]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA327654EA 1050 EUR
ORIGTA327654
Anti-CD79A Mouse Monoclonal Antibody [clone: JCB117]
Anticorps
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: CD79A
Clonality: Monoclonal
Clone: JCB117
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR