Imprimer

Anti-GABAA Rabbit Polyclonal Antibody

Fournisseur: Cayman Chemical
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

CAYM10600-1EA 650 EUR
CAYM10600-1
Anti-GABAA Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: GABAA
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR