Imprimer
Anti-IgY Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CAYM13413-1EA 191 EUR
CAYM13413-1
Anti-IgY Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Anticorps
Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: R-PE (Phycoerythrin) (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype:
Reactivity: Chicken