Imprimer
Blood sedimentation pipette, acc. to Westergren
Fournisseur: Marienfeld
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MARI3268062EA 42.2 EUR
MARI3268062
Blood sedimentation pipette, acc. to Westergren
Pipettes Pipettes pour hémomètres
In case of suspected infectious diseases the erythrocyte sedimentation rate of blood is examined by means of sedimentation pipettes according to Westergren.
  • Clear AR-Glas®
  • White graduation 0 to 200 in 1 mm
  • For micro method
Suitable for in-vitro diagnostic (IVD) applications according to IVD directive 98/79/EC.