Imprimer

Anti-NCAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM 188]

Fournisseur: Abnova
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABNOMAB6001EA 399 EUR
ABNOMAB6001
Anti-NCAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM 188]
Anticorps
Mouse monoclonal antibody raised against NCAM1.

Recommended Dilutions: Flow Cytometry: 10 ?l/10^6 cells

Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: MEM 188
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR