Imprimer

Anti-ABI1 Rabbit Polyclonal Antibody

Fournisseur: Abnova
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABNOPAB5221EA 399 EUR
ABNOPAB5221
Anti-ABI1 Rabbit Polyclonal Antibody
Anticorps
Rabbit polyclonal antibody raised against synthetic peptide of ABI1.

Type: Primary
Antigen: ABI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR