Imprimer

Anti-MS4A1 Mouse Monoclonal Antibody (Spectral Red®) [clone: B-Ly1]

Fournisseur: Abnova
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABNOMAB5994EA 419 EUR
ABNOMAB5994
Anti-MS4A1 Mouse Monoclonal Antibody (Spectral Red®) [clone: B-Ly1]
Anticorps
Mouse monoclonal antibody raised against MS4A1.

Recommended Dilutions: Flow Cytometry: 10 ?l/10^6 cells

Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: B-Ly1
Conjugation: Spectral Red®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR