Imprimer

Anti-CD19 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: SJ25-C1]

Fournisseur: Abnova
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABNOMAB5988EA 518 EUR
ABNOMAB5988
Anti-CD19 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: SJ25-C1]
Anticorps
Mouse monoclonal antibody raised against native CD19.

Recommended Dilutions: Flow Cytometry: 10 ?l/10^6 cells

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25-C1
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR