Imprimer

Anti-RRBP1 Rabbit Polyclonal Antibody

Fournisseur: Abnova
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABNOPAB15696EA 643 EUR
ABNOPAB15696
Anti-RRBP1 Rabbit Polyclonal Antibody
Anticorps
Rabbit polyclonal antibody raised against partial recombinant Rrbp1.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: RRBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR