Imprimer

Anti-UNQ1887 Rabbit Polyclonal Antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABNOPAB4380EA 351 EUR
ABNOPAB4380
Anti-UNQ1887 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic peptide of UNQ1887.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: UNQ1887
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR