Imprimer

Anti-ITGAE Mouse Monoclonal Antibody [clone: LF61]

Fournisseur: Abnova
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABNOMAB5260EA 643 EUR
ABNOMAB5260
Anti-ITGAE Mouse Monoclonal Antibody [clone: LF61]
Anticorps
Mouse monoclonal antibody raised against native ITGAE.

Type: Primary
Antigen: ITGAE
Clonality: Monoclonal
Clone: LF61
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR