Imprimer

Anti-Myelin Mouse Monoclonal Antibody [clone: 2B5]

Fournisseur: Abnova
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABNOMAB5259EA 518 EUR
ABNOMAB5259
Anti-Myelin Mouse Monoclonal Antibody [clone: 2B5]
Anticorps
Mouse monoclonal antibody raised against native Myelin.

Recommended Dilutions: Immunohistochemistry: 1:10

Type: Primary
Antigen: Myelin
Clonality: Monoclonal
Clone: 2B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR