Imprimer

Anti-ARR3 Goat Polyclonal Antibody

Fournisseur: Abnova
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABNOPAB15683EA 518 EUR
ABNOPAB15683
Anti-ARR3 Goat Polyclonal Antibody
Anticorps
Goat polyclonal antibody raised aganist synthetic peptide of ARR3.

Recommended Dilutions: ELISA: 1:1000

Type: Primary
Antigen: ARR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR