Imprimer

Anti-GP5 Mouse Monoclonal Antibody [clone: CLB-SW16]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABNOMAB5254EA 1150 EUR
ABNOMAB5254
Anti-GP5 Mouse Monoclonal Antibody [clone: CLB-SW16]
Mouse monoclonal antibody raised against native GP5.

Type: Primary
Antigen: GP5
Clonality: Monoclonal
Clone: CLB-SW16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR