Imprimer

Anti-MED17 Rabbit Polyclonal Antibody

Fournisseur: Abnova
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABNOPAB1164EA 364 EUR
ABNOPAB1164
Anti-MED17 Rabbit Polyclonal Antibody
Anticorps
Rabbit polyclonal antibody raised against full length recombinant Saccharomyces cerevisiae SRB4.

Type: Primary
Antigen: MED17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR