Imprimer
Breakup solution
Fournisseur: SureScreen Diagnostics
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SUREBUSEA 0 EUR
SUREBUS
Breakup solution
Tests pour le dépistage de drogue
For use with raw drug testing kits. This solution is used to break up organic material, such as cannabis leaf, prior to running a drug test using the fluid as sample.