Imprimer
Pregnancy test
Fournisseur: SureScreen Diagnostics
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SUREGHCGBCEA 39.6 EUR
SUREGHCGBC
Pregnancy test
Systèmes de diagnostic clinique Clinical Reagents and Kits
Human chorionic gonadotropin (hCG) is a glycoprotein hormone produced by the developing placenta shortly after fertilisation in normal pregnancy.
  • Easy to perform routine test
  • Gives results in three minutes only
  • hCG can be detected in both urine and serum 7 to 10 days after conception
These test strips are the industry-standard for pregnancy testing.
Cdt: Sealed in a foil pouch, which ensures the quality of the product throughout the shelf-life.