Imprimer
Transfection reagent kit, NanoJuice®
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
71902-4EA 883 EUR
NOVG71902-4
Transfection reagent kit, NanoJuice®
The NanoJuice™ Transfection Kit is designed for stable and transient transfections of primary cells, Saos-2, Caco-2, HUVEC, Jurkat, Raw 264.7, HeLa, and other mammalian cells. Complete control over transfection conditions allows optimisation of transfection efficiencies and reduces cytotoxicity. The NanoJuice™ Transfection Kit provides flexibility to fine tune transfection conditions for efficient DNA delivery.
  • Minimal cellular toxicity
  • Derived from non animal sources
  • Eliminates the need for media changes
  • Flexible reaction conditions to optimise for specific cell types
  • Compatible with both serum-containing and serum-free media
Storage temperature: +2 to +8 °C.
Informations de livraison: The NanoJuice™ Transfection Kit provides enough components to perform 240 transfections in ten 24-well plates or 2400 transfections in one hundred 24-well plates. The NanoJuice™ Core Transfection Reagent and the NanoJuice™ Transfection Booster are supplied as ready-to-use sterile solutions.