Imprimer
Transfection booster, NanoJuice®
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
71901-3EA 308 EUR
NOVG71901-3
Transfection booster, NanoJuice®
NanoJuice™ transfection booster is designed to be used in concert with the NanoJuice™ Core transfection reagent.
Storage temperature: +2 to +8°C.
Informations de livraison: Available separately in a 1 ml volume for greater flexibility and experimental scale-up.