Imprimer
Cryopreservation media, ProFreeze™-CDM
Fournisseur: Lonza
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BioWhittaker®
LONZ12-769EEA 176 EUR
LONZ12-769E
Cryopreservation media, ProFreeze™-CDM
ProFreeze™-CDM is a chemically defined freeze medium suitable for cryopreserving many cell types in the absence of foetal bovine serum (FBS).
  • Non animal, protein-free medium
  • Maintains high cell viability upon recovery from frozen storage
Requires addition of DMSO at the time of use.
Storage temperature: 2 to 8 °C