Imprimer
Cross cut adhesion test kits, CC2000
Fournisseur: Industrial Physics
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SIMESP1690EA 406 EUR
SIMESP1690
Cross cut adhesion test kits, CC2000
Testeurs d'adhérence
This Cross Cut adhesion test kit (CC2000) is used to test the adhesion of dry coats of paint on their substrate by means of a series of cuts through the coating.
  • Self-adjusting knife-holder ensures equal pressure on the cutting knife
  • Ergonomically shaped handle
  • Easy to change cutting knife, no extra key needed
  • Wide range of knife sizes available for different coating thicknesses and substrates and according to different standards
Two series of parallel cuts cross angled to each other to obtain a pattern of 25 or 100 similar squares. The ruled area is evaluated by using a table chart after a short treatment with a stiff brush, or adhesive tape for hard substrates.
Certifications: EN ISO 2409, ASTM D3359
Informations de livraison: Kit includes CC2000 grip, cutter (type varies), brush, illuminated loupe and roll of adhesive tape acc. EN-ISO 2409