Imprimer
Flow meters for water
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
GLPR10720EA 143 EUR
GLPR10720 GLPR10719 GLPR10718 GLPR10726 GLPR10725 GLPR10717 GLPR10727 GLPR10722 GLPR10724 GLPR10723
Flow meters for water
Borosilicate glass tubes with polyethylene connectors, linear or direct reading scale giving water flow measurements. Clear indelibly fired-in markings and yellow backing ensure easy reading.
  • Repeatability of ±0,25 % of full scale reading
  • Rib guided flow tube for maximum stability and accuracy
  • Heat-resistant glass with heavy walls and push-on polythene connectors