Garder ma session ouverte ?
Fin de session dans  
La session a expirée
Votre session a expiré. Pour votre sécurité, nous vous avons déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-S100A8 Mouse Monoclonal Antibody [clone: IMG48M7C7]

Marques : BioVision
S100A8 is a low-molecular weight member of the S100 family of calcium-binding protein which promotes tumorigenesis. It promotes cell migration and invasion through p38 MAP dependent NF-kappaB activation leading to an increase of MMP2 and MMP12 in gastric cancer. The phagocyte-specific Ca2+-binding S100A8 protein has been proposed as an essential regulator of the plasma membrane NADPH oxidase activity. It is abundantly expressed in the cytosol of neutrophils and is able to form Ca2+-dependent heterocomplexes, with heterotetramers being a probable prerequisite for its biological activities in myeloid cells. S100A8 and S100A9 have been proposed as essential regulators that exert their role through interactions with NADPH oxidase subunits. S100A8 and S100A9 are generally considered proinflammatory. Whereas hypohalous acids generated by activated phagocytes promote novel modifications in murine S100A8 but modifications to human S100A8 are undefined and there is no evidence that these proteins scavenge oxidants in human disease. Oxidized S100A8 was prominent in lungs from patients with asthma and significantly elevated in sputum compared to controls. Results have broad implications for conditions under which hypohalous acid oxidants are generated by activated phagocytes. Identification in human disease of the novel S100A8 Cys derivatives typical of those generated in vitro strongly supports the notion that S100A8 contributes to antioxidant defense during oxidative stress.

Recommended Dilutions: IHC (P): use antibody at 2 µg/ml, Western blot analysis: 0.5-5 µg/ml.

Type: Primary
Antigen: S100A8
Clonality: Monoclonal
Clone: IMG48M7C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse



 

COMMANDE

Développer tout / Réduire tout
Taille Cdt Type de conditionnement Référence VWR Quantité
100 μg 100 µG Vial BIOV6196-100
Generate Barcodes Barcode Label Format: 
Créer les pages catalogue sous PDF

SPÉCIFICATIONS

Symbole antigène S100A8
Nom de l'antigène S100 calcium binding protein A8
Réactivité Mouse, Human
Conjugaison Unconjugated
Clonalité Monoclonal
Type d' Anticorps Primary
Clone IMG48M7C7
Hôte Mouse
Isotype IgG2b
Western blot Yes
ImmunoChemistry Yes
Antigen synonyms MIF, CP-10, MRP8, P8, 60B8AG, MA387, CGLA, NIF, CFAG, CAGA, L1Ag
Gene ID 6279
Mesures de concentration 0,5 mg/ml
Cross adsorption No
Immunogen Amino acids 2-93 of human S100A8
Purification Protein G purified
Form Liquid
Storage buffer 0,2 ml PBS containing 0,05% BSA and 0,05% sodium azide.
Température de stockage – 20 °C
Shipping temperature 4 °C

ÉCRIRE UN AVIS

Besoin d'aide ?

  • Précisez ce que vous avez aimé et ce que vous n'avez pas apprécié.
  • Le produit répond-il à vos attentes ?
  • A qui le recommanderiez-vous ?

C'est sous ce nom que VWR va publier votre avis.
Nous recommandons aux auteurs d'utiliser leurs initiales.

Merci de confirmer votre secteur d'activité

Exemple : Recherche Pharmaceutique

Merci de confirmer votre fonction

Exemple : Chercheur

Merci de confirmer votre ville

Exemple : Lille, Clermont

AVIS (0)

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Classer par:

Entrer les détails pour la recherche

Sélectionner les article pour lesquels vous recherchez le certificat

Indiquer le N° de lot. Ce champs est obligatoire. Entrer un "X" si inconnu pour les PC.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Ou
Editer et comparer les propositions
Nous avons trouvé des articles alternatifs qui vous permettraient d économiser jusque par emballage :   Pour comparer selectionnez jusqu à 3 propositions ci-dessous et cliquer sur comparer. Pour ajouter des articles dans votre panier, entrez une quantité et cliquez sur Ajoutez au panier.

Produit initial

Description Référence VWR Disponibilité Unité Votre prix Prix par Quantité
 

Un nouvel aspect et un engagement renouvelé

Axée sur les fonctionnalités et les ressources que vous connaissez, optimisée avec l’innovation Avantor®. Notre nouvelle marque est synonyme d’agilité et de capacité d’adaptation aux besoins de nos clients.

EN SAVOIR PLUS COMMENCEZ VOS ACHATS