Imprimer

Anti-Ferredoxin Reductase Mouse Monoclonal Antibody [clone: 6C2]

Fournisseur: BioVision
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BIOV6216-100EA 367 EUR
BIOV6216-100
Anti-Ferredoxin Reductase Mouse Monoclonal Antibody [clone: 6C2]
Ferredoxin reductase is a ubiquitous flavoenzyme, containing noncovalently bound FAD as a prosthetic group. It plays a role in delivering NADPH or low potential one-electron donors such as ferredoxin and flavodoxin to redox-based metabolisms in plastids, mitochondria and bacteria. In mammals, ferredoxin reductase is loosely associated with the inner mitochondrial membrane and receives electrons from NADPH. These electrons are transferred to ferredoxin which shuttles electrons to cytochrome P450 in the adrenal cortex mitochondrial steroid hydroxylation systems. It serves as the first electron transfer protein in all the mitochondrial P450 systems. Including cholesterol side chain cleavage in all steroidogenic tissues, steroid 11-beta hydroxylation in the adrenal cortex, 25-OH-vitamin D3-24 hydroxylation in the kidney, and sterol C-27 hydroxylation in the liver.

Recommended Dilutions: WB

Type: Primary
Antigen: Ferredoxin Reductase
Clonality: Monoclonal
Clone: 6C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR