Imprimer
Anti-HDAC3 Rabbit Polyclonal Antibody
Fournisseur: BioVision
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BIOV6603-100EA 299 EUR
BIOV6603-100
Anti-HDAC3 Rabbit Polyclonal Antibody
HDAC 3 is a member of the class I mammalian histone deacetylase family, which plays an important role in modulating the eukaryotic chromatin structure. Human HDAC 3 is composed of 428 amino acid residues. HDAC 3 interacts with a growing number of transcriptional factors including SMRT and N-CoR. The interacting complexes bind to specific regions of chromatin and regulate gene transcription in these regions.
Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 20-30 µg/ml)
Type: Primary
Antigen: HDAC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat