Imprimer

Anti-Adipsin Rabbit Polyclonal Antibody

Fournisseur: BioVision
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BIOV3937-100EA 276 EUR
BIOV3937-100
Anti-Adipsin Rabbit Polyclonal Antibody
Adipsin is a serine protease secreted by adipose tissue, and it has also been found in monocytes/macrophages. Adipsin has been identified to be similar to complement Factor D. Adipsin cleaved complement Factor B when the later form complexed with activated complement component C3. Low level expression of Adipsin has been linked to obesity in mouse model sμggesting that defects in Adipsin is the result of this systemic disorder.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Adipsin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR