Imprimer

Anti-LPL Rabbit Polyclonal Antibody

Fournisseur: BioVision
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BIOV3947-100EA 305 EUR
BIOV3947-100
Anti-LPL Rabbit Polyclonal Antibody
Lipoprotein lipase (LPL) is mainly expressed in the heart, muscle and adipose tissue. LPL is a homodimer that mediates the hydrolysis of triglycerides of very low density lipoproteins and also function as ligand/bridging factor for receptor-mediated lipoprotein uptake. Defects in LPL may result in hypertriglceridemia.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: LPL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR