Imprimer

Anti-Thy1/CD90 Rabbit Polyclonal Antibody

Fournisseur: BioVision
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BIOV6778-100EA 276 EUR
BIOV6778-100
Anti-Thy1/CD90 Rabbit Polyclonal Antibody
Thy1 is a GPI-anchored, developmentally regulated protein involved in various signaling cascades that mediate neurite outgrowth, T cell activation, tumor suppression, apoptosis, and fibrosis. It is highly expressed on the surface of adult neurons and is thought to play a role in modulating adhesive and migratory events, such as neurite extension. Decreased Thy1 expression is associated with the development of epithelial ovarian cancer, revealing its role as a putative tumor suppressor gene of human ovarian cancer. Thy1 knockout mice have impaired cutaneous immune responses and abnormal retinal development. Thy1 is epigenetically regulated or deregulated in some disease states, such as pulmonary fibrosis. The potentially reversible hypermethylation of the Thy1 promoter offers the possibility of novel therapeutic options in this disease.

Recommended Dilutions: Western Blot: 1:1000, IHC: 1:50-1:100.

Type: Primary
Antigen: Thy1/CD90
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat.
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR