Optical oxygen sensor, FDO® 1100 IDS
Supplier: SI Analytics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IDS®
SCOI285202440EA 1330 GBP
SCOI285202440
Optical oxygen sensor, FDO® 1100 IDS
Electrodes Dissolved Oxygen Electrodes
Optical oxygen measuring electrode with plastic shaft and temperature sensor NTC 30 kOhm for versatile applications.
  • Intelligent membrane (IQMC technology): Individual calibration values are determined by a factory calibration process and stored on the memory chip of the sensor cap
  • Automatic sensor recognition: Data of the sensor and sensor cap is recalled by the meter when the sensor is connected
  • Digital transmission technique guarantees the failure-free communication with the meter even with long connection cables
A memory chip is integrated in the sensor cap. The following data are stored on the memory chip:
- Type designation of the sensor cap
- Series number
- Data of the factory calibration or user calibration
Recommended fields of application: On site measurements in rivers, lakes and wastewater, applications in water laboratories, BOD measurements
Delivery information: Supplied with sensor cap, check and storage vessel and operating manual.