Imprimer
Luxmeter, digital, LU8500
Fournisseur: Industrial Physics
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SIMELU8500EA 392 EUR
SIMELU8500
Luxmeter, digital, LU8500
LU8500 luxmeter is a digital, handheld with large LCD display.
  • High accuracy and rapid response
  • Peak-hold function for tracing peak signals of light pulse with least duration 10 µs
  • Capable of selecting measuring mode in lux or fc scale alternatively
  • Measurement rate: 1,5 times per second
  • With maximum and minimum measurements
Informations de livraison: The instrument is delivered with a carrying case, instruction manual and battery.