Imprimer
Votre recherche pour: Billes magnétiques
29  résultat(s)
SearchResultCount:"29"
Sort Results
Vue liste Nouvelle Vue
Evaluer les résultats de votre recherche
Fournisseur: Merck
Description: Silica particles, MagPrep®
Référence Produit: (1.01899.0050)
Fournisseur: Merck
Description: MagPrep® Silica HS beads
UOM: 1 * 50 mL
Référence Produit: (ROCKTMS-15-50)
Fournisseur: Rockland Immunochemicals
Description: 6x1,5 ml Position magnetic separator
UOM: 1 * 1 Pce
Fournisseur: Thermo Fisher Scientific
Description: Protein A/G magnetic agarose beads provide a fast, convenient method for purification of immunoglobulins from serum, cell culture supernatant, or asci...
Fournisseur: Thermo Fisher Scientific
Description: Magnetic beads are a convenient affinity support for a variety of assays, which allow easy purification of the target without columns or centrifugatio...
Fournisseur: Thermo Fisher Scientific
Description: The Pierce™ Ni-NTA magnetic beads are high-capacity nickel-IMAC beads for affinity purification of His-tagged fusion proteins in manual or automated f...
Fournisseur: Thermo Fisher Scientific
Description: The Pierce™ Anti-c-Myc magnetic beads are affinity particles for immunoprecipitation of recombinant c-Myc-tagged proteins expressed in bacterial or ma...
Fournisseur: Thermo Fisher Scientific
Description: These protein L magnetic beads are high-capacity and high-throughput affinity particles for antibody purification using manual or robotic magnetic sep...
Fournisseur: Thermo Fisher Scientific
Description: These Pierce™ streptavidin magnetic beads accelerate throughput for automated magnetic purification of biotinylated molecules.
Fournisseur: Thermo Fisher Scientific
Description: These protein magnetic beads are high-capacity and high-throughput affinity particles for antibody purification and immunoprecipitation methods using ...
Fournisseur: Thermo Fisher Scientific
Description: The Pierce™ NHS-activated magnetic beads enable covalent, amine-based conjugation of proteins to magnetic beads in a simple mix-and-go format for use ...
Fournisseur: Thermo Fisher Scientific
Description: Pierce™ protein A/G magnetic beads are high-performance affinity particles for antibody purification and immunoprecipitation methods using manual or r...
Fournisseur: Thermo Fisher Scientific
Description: The Pierce™ Anti-HA magnetic beads are affinity particles for immunoprecipitation of recombinant HA-tagged proteins expressed in bacterial or mammalia...
Référence Produit: (PIER78605)
Fournisseur: Thermo Fisher Scientific
Description: These magnetic agarose beads consist of highly crosslinked agarose beads embedded with magnetite and a covalently attached tetradentate nitrilotriacet...
UOM: 1 * 4 mL
Fournisseur: Cytiva
Description: NHS Mag Sepharose magnetic beads are designed for pull-down techniques, involving isolation of a protein through adsorption of complex samples onto be...
Fournisseur: Cytiva
Description: Sera-Mag Carboxylate-Modified Magnetic Beads & SpeedBeads have free carboxyl groups on the surface for convenient covalent coupling of target molecule...
Prix sur demande
Le stock de cet article est limité mais peut être disponible dans un entrepôt proche de vous. Merci de vous assurer que vous êtes connecté sur le site afin que le stock disponible soit affiché. Si l'call est toujours affiché et vous avez besoin d'aide, s'il vous plaît appelez-nous au 01 45 14 89 12.
Le stock de cet article est limité mais peut être disponible dans un entrepôt proche de vous. Merci de vous assurer que vous êtes connecté sur le site afin que le stock disponible soit affiché. Si l'call est toujours affiché et vous avez besoin d'aide, s'il vous plaît appelez-nous au 01 45 14 89 12.
Ces articles ne peuvent pas être ajoutés à votre panier en raison des exigences réglementaires. Veuillez envoyer un e-mail à webshop.fr@vwr.com
Veuillez noter qu'une autorisation peut être nécessaire pour commander ce produit. Un représentant de VWR vous contactera si votre commande le nécessite.
Ce produit a été bloqué par votre entreprise. Veuillez s'il vous plait contacter votre service achats pour plus d'informations.
Le produit original n'est plus disponible. Le remplacement représenté est disponible
Ce produit n'est plus disponible. Des alternatives peuvent être disponibles en faisant une recherche avec la référence VWR ci-dessus. Si vous avez besoin d'aide, s'il vous plaît appelez le Service Clients au 0825 02 30 30 (0,18 € TTC/mn).
1 - 16 of 29
no targeter for Bottom