Imprimer
Wound cleansers and plasters, Salvequick
Fournisseur: Orkla
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Salvequick
CEDE1880EA 243 EUR
CEDE1880
Wound cleansers and plasters, Salvequick
Matériel de premiers secours Pansements
Wound cleansers and adhesive bandages (plasters) packed in a PE plastic zip bag. The wound cleansers are intended for mechanical cleansing of minor wounds. The adhesive bandages are intended to be used as mechanical barrier and for absorption of exudates from minor wounds.
  • Wound cleansers: Sterile tissues soaked in 0,9% NaCl
  • Adhesive bandages: Micro perforated PE foil coated with polyacrylate adhesive; non woven wound pad (viscose and PP/PE) with a non stick wound contact layer (PE); wrapped in cold seal paper
Adhesive bandages: 25×72 mm