Imprimer
Ion selective electrode, half-cell, sulphide
Fournisseur: Hach
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HACHE41M016EA 3410 EUR
HACHE41M016
Ion selective electrode, half-cell, sulphide
Électrodes Électrodes ISE
The ISE25S-9 is a half-cell ISE and requires an external reference.
  • Plastic body, non refillable internal electrolyte, and sulfide (S-) or silver (Ag+) ion specific solid-state sensor
  • Factory controlled to ensure high quality and premium performance
  • Sturdy structural properties
Compatible with sensION+ benchtop meters (or any ion meter with a BNC connection).
Informations de livraison: Supplied with Certificate of Conformity. The electrode is supplied without cable which needs to be ordered separately.