Imprimer
Ion selective electrodes, chloride, IntelliCAL™
Fournisseur: Hach
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
IntelliCAL™
HACHISECL18103EA 1510 EUR
HACHISECL18103 HACHISECL18101 HACHISECL181AP
Ion selective electrodes, chloride, IntelliCAL™
Électrodes Électrodes ISE
IntelliCAL™ ISECL181 is a digital, combination chloride ion selective electrode (ISE) with a non refillable, double junction reference and built-in temperature sensor. The solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE.
  • Compatible with digital Hach meters only (e.g. HQd meters)
  • Requires virtually no maintenance
  • Fast, stable, and accurate results
  • Ultimate traceability in measurement history
  • Can be moved between meters without the need to re-calibrate or re-enter measurement settings
The probe is available with a 1 or 3 meter cable and is intended for laboratory use. Ideal for measuring chloride concentrations in wastewater, drinking water and general water quality applications.
Informations de commande: Electrode is supplied with calibration certificate and basic user manual.
The analysis package contains: Electrode IntelliCAL™ ISECL18101, chloride ionic strength adjustor powder pillows (100), chloride 100 mg/l standard solution (1 l), and chloride 1000 mg/l standard solution (500 ml).