Imprimer
Western blotting filter paper, extra thick, Pierce™
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PIER88605EA 75 EUR
PIER88605 PIER88615 PIER88610 PIER88620
Western blotting filter paper, extra thick, Pierce™
Papiers buvard
These Western blotting filter papers are pre-cut cotton sheets for wet or semi-dry, passive or electrophoretic transfer of proteins from polyacrylamide gels (SDS-PAGE) to PVDF, nitrocellulose, or other membranes.
  • Extra-thick filter paper for high capacity blotting or to eliminate using multiple sheets
  • Pre-cut filter paper sheets in several convenient sizes for use with most mini gel sizes, tank transfer cassettes and semi-dry blotters
  • Tested and recommmend for use with various protein methods, including tank and semi-dry transfer
Clean cotton cellulose fibre paper manufactured with additive-free ultrapure water to eliminate sources of background signal and artifacts.